Gene Transcripts

Gene Symbol
Tbl1xr1
Entrez Gene ID
365755
Gene Name
transducin (beta)-like 1 X-linked receptor 1
Gene Aliases
RGD1560053
Chromosome Location
Chr. 2: 107221987 - 107359229 on Build Rnor_6.0
UniGene ID
Rn.140831
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108941.1 NP_001102411.1 13 1361
XM_006232190.2 XP_006232252.1 12 1254
XM_008760889.1 XP_008759111.1 13 2117

View Full Product Details