Gene Transcripts

Gene Symbol
Krt5
Entrez Gene ID
369017
Gene Name
keratin 5
Gene Aliases
Krt2-5, Sak57
Chromosome Location
Chr. 7: 143320142 - 143324536 on Build Rnor_6.0
UniGene ID
Rn.129725
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183333.1 NP_899162.1 1678
GenBank mRNA AY342389.1 - 1678
BC062086.1 - 1541 5599969

View Full Product Details