Gene Transcripts
- Gene Symbol
- Rmi2
- Entrez Gene ID
- 497856
- Gene Name
- RecQ mediated genome instability 2
- Gene Aliases
- RGD1563163
- Chromosome Location
- Chr. 10: 4904458 - 4910339 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_573037.4 |
XP_573037.1
|
1, 2 |
343 |
|
View Full Product Details