Gene Transcripts

Gene Symbol
Trmt13
Entrez Gene ID
499697
Gene Name
tRNA methyltransferase 13 homolog (S. cerevisiae)
Gene Aliases
Ccdc76
Chromosome Location
Chr. 2: 219615665 - 219628824 on Build Rnor_6.0
UniGene ID
Rn.154930
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033902.2 NP_001029074.2 1, 2 179
GenBank mRNA BC103656.1 - 1, 2 227
BC166503.1 - 1, 2 179 7122476

View Full Product Details