Gene Transcripts

Gene Symbol
Btbd2
Entrez Gene ID
500793
Gene Name
BTB (POZ) domain containing 2
Gene Aliases
RGD1566094
Chromosome Location
Chr. 7: 11929891 - 11943559 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006241037.2 XP_006241099.2 3 547
XM_008765152.1 XP_008763374.1 3 547

View Full Product Details