Gene Transcripts

Gene Symbol
Lrrc19
Entrez Gene ID
680478
Gene Name
leucine rich repeat containing 19
Gene Aliases
Chromosome Location
Chr. 5: 113615772 - 113626832 on Build Rnor_6.0
UniGene ID
Rn.178166
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109413.2 NP_001102883.1 4 905
XM_006238402.2 XP_006238464.1 5 1267

View Full Product Details