Gene Transcripts

Gene Symbol
Fpr-rs6
Entrez Gene ID
690230
Gene Name
formyl peptide receptor, related sequence 6
Gene Aliases
Chromosome Location
Chr. 1: 86262503 - 86263534 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001073753.1 XP_001073753.1 1 101

View Full Product Details