Gene Transcripts

Gene Symbol
NCAPD2
Entrez Gene ID
9918
Gene Name
non-SMC condensin I complex subunit D2
Gene Aliases
CAP-D2, CNAP1, hCAP-D2
Chromosome Location
Chr. 12: 6494132 - 6531966 on Build GRCh38
UniGene ID
Hs.5719
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014865.3 NP_055680.3 29 3775
GenBank mRNA AK022511.1 - 29 3775
AK128354.1 - 29 3687
AK303310.1 - 28 3750
BC028182.1 - 29 3787 5395775
D63880.1 - 29 4528

View Full Product Details