Gene Transcripts
- Gene Symbol
- TNFSF12-TNFSF13
- Entrez Gene ID
- 407977
- Gene Name
- TNFSF12-TNFSF13 readthrough
- Gene Aliases
- TWE-PRIL
- Chromosome Location
- Chr. 17: 7549058 - 7561608 on Build GRCh38
- UniGene ID
- Hs.54673
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_172089.3 |
NP_742086.1
|
6, 7 |
590 |
|
GenBank mRNA |
AY081051.1 |
- |
6, 7 |
549 |
|
View Full Product Details