Gene Transcripts

Gene Symbol
SLC35G5
Entrez Gene ID
83650
Gene Name
solute carrier family 35 member G5
Gene Aliases
AMAC, AMAC1L2
Chromosome Location
Chr. 8: 11330986 - 11332186 on Build GRCh38
UniGene ID
Hs.458397
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_054028.1 NP_473369.1 1 1091
GenBank mRNA AJ291677.1 - 1 1091
BC131696.1 - 1 1058 40120574

Gene Symbol
SLC35G4
Entrez Gene ID
646000
Gene Name
solute carrier family 35 member G4
Gene Aliases
AMAC1L1, SLC35G4P
Chromosome Location
Chr. 18: 11609558 - 11610612 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282300.1 NP_001269229.1 1 1008

View Full Product Details