Gene Transcripts

Gene Symbol
KLHL17
Entrez Gene ID
339451
Gene Name
kelch like family member 17
Gene Aliases
Chromosome Location
Chr. 1: 960103 - 965719 on Build GRCh38
UniGene ID
Hs.109212
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006710600.3 XP_006710663.1 1 104
XM_006710601.3 XP_006710664.1 1 104

View Full Product Details