Gene Transcripts

Gene Symbol
Slc51b
Entrez Gene ID
330962
Gene Name
solute carrier family 51, beta subunit
Gene Aliases
Ostb
Chromosome Location
Chr. 9: 65412753 - 65422773 on Build GRCm38
UniGene ID
Mm.335247
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178933.2 NP_849264.1 4 257
GenBank mRNA AK002463.1 - 4 388
AK004188.1 - 4 440
AY279396.1 - 4 257
BC115607.1 - 3 236 40102691

View Full Product Details