Gene Transcripts
- Gene Symbol
- EPHA4
- Entrez Gene ID
- 2043
- Gene Name
- EPH receptor A4
- Gene Aliases
- HEK8, SEK, TYRO1
- Chromosome Location
- Chr. 2: 221418027 - 221574202 on Build GRCh38
- UniGene ID
- Hs.371218
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001304536.1 |
NP_001291465.1
|
10, 11 |
2050 |
|
NM_001304537.1 |
NP_001291466.1
|
8, 9 |
1817 |
|
NM_004438.4 |
NP_004429.1
|
9, 10 |
1885 |
|
XM_005246374.2 |
XP_005246431.1
|
9, 10 |
1885 |
|
GenBank mRNA |
AK290306.1 |
- |
10, 11 |
2050 |
|
AK300772.1 |
- |
8, 9 |
1755 |
|
AK312380.1 |
- |
9, 10 |
1831 |
|
BC026327.1 |
- |
9, 10 |
1810 |
4814429 |
BC105000.1 |
- |
9, 10 |
1811 |
8144003 |
BC105002.1 |
- |
9, 10 |
1811 |
8144005 |
L36645.1 |
- |
9, 10 |
1808 |
|
View Full Product Details