Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001271440.1 | NP_001258369.1 | 7 | 1316 | |
NM_001271441.1 | NP_001258370.1 | 7 | 1676 | ||
NM_001271442.1 | NP_001258371.1 | 8 | 1224 | ||
NM_004928.2 | NP_004919.1 | 7 | 1319 | ||
XM_006724051.3 | XP_006724114.1 | 6 | 1448 | ||
XM_006724053.3 | XP_006724116.1 | 6 | 1125 | ||
XM_017028470.1 | XP_016883959.1 | 7 | 1327 | ||
XM_017028471.1 | XP_016883960.1 | 6 | 1149 | ||
XM_017028472.1 | XP_016883961.1 | 6 | 1204 | ||
GenBank mRNA | Y11392.1 | - | 7 | 1179 | |
Z93322.1 | - | 7 | 1118 |