Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005052.2 | NP_005043.1 | 5, 6 | 554 | |
GenBank mRNA | AF008591.1 | - | 5, 6 | 518 | |
AF498966.1 | - | 5, 6 | 448 | ||
BC009605.1 | - | 5, 6 | 555 | 4284122 | |
BC015197.1 | - | 5, 6 | 555 | 4275237 | |
BT019443.1 | - | 5, 6 | 448 |