Gene Transcripts

Gene Symbol
KRTAP19-6
Entrez Gene ID
337973
Gene Name
keratin associated protein 19-6
Gene Aliases
KAP19.6
Chromosome Location
Chr. 21: 30541535 - 30541864 on Build GRCh38
UniGene ID
Hs.553695
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303120.1 NP_001290049.1 1 176
GenBank mRNA AB096947.1 - 1 174
AB096948.1 - 1 174
BC101813.1 - 1 176
BC101815.1 - 1 176

View Full Product Details