Gene Transcripts
- Gene Symbol
- FASTKD1
- Entrez Gene ID
- 79675
- Gene Name
- FAST kinase domains 1
- Gene Aliases
- Chromosome Location
- Chr. 2: 169527879 - 169573921 on Build GRCh38
- UniGene ID
- Hs.529276
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001281476.2 |
NP_001268405.1
|
6, 7 |
1477 |
|
NM_001322046.1 |
NP_001308975.1
|
6, 7 |
2150 |
|
NM_001322048.1 |
NP_001308977.1
|
5, 6 |
1405 |
|
NM_001322049.1 |
NP_001308978.1
|
5, 6 |
1408 |
|
NM_024622.5 |
NP_078898.3
|
6, 7 |
1477 |
|
NR_104020.2 |
|
6, 7 |
1477 |
|
XM_006712750.3 |
XP_006712813.1
|
5, 6 |
1403 |
|
XM_006712751.3 |
XP_006712814.1
|
2, 3 |
1488 |
|
XM_017004911.1 |
XP_016860400.1
|
6, 7 |
2790 |
|
XM_017004912.1 |
XP_016860401.1
|
2, 3 |
1488 |
|
GenBank mRNA |
AB058703.1 |
- |
4, 5 |
659 |
|
AK025554.1 |
- |
3, 4 |
589 |
|
AK055892.1 |
- |
6, 7 |
1388 |
|
AK074302.1 |
- |
6, 7 |
1368 |
|
AK130797.1 |
- |
6, 7 |
1400 |
|
AK225293.1 |
- |
3, 4 |
589 |
|
AL832058.1 |
- |
6, 7 |
1360 |
|
BC032687.2 |
- |
5, 6 |
806 |
5581443 |
View Full Product Details