Gene Transcripts
- Gene Symbol
- Speer4d
- Entrez Gene ID
- 360220
- Gene Name
- spermatogenesis associated glutamate (E)-rich protein 4d
- Gene Aliases
- 4933431D05Rik, SPEER-4D
- Chromosome Location
- Chr. 5: 15619099 - 15623864 on Build GRCm38
- UniGene ID
- Mm.425235
- Species
- Mus musculus
- Gene Symbol
- 4930572O03Rik
- Entrez Gene ID
- 381715
- Gene Name
- spermatogenesis associated glutamate (E)-rich protein pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 5: 15652288 - 15657059 on Build GRCm38
- UniGene ID
- Mm.382859
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK076955.1 |
- |
5 |
1004 |
|
- Gene Symbol
- Speer4c
- Entrez Gene ID
- 100502967
- Gene Name
- spermatogenesis associated glutamate (E)-rich protein 4c
- Gene Aliases
- 4933431D05Rik, SPEER-4C
- Chromosome Location
- Chr. 5: 15709505 - 15714675 on Build GRCm38
- UniGene ID
- Mm.425235
- Species
- Mus musculus
View Full Product Details