Gene Transcripts

Gene Symbol
Zglp1
Entrez Gene ID
100009600
Gene Name
zinc finger, GATA-like protein 1
Gene Aliases
Glp1
Chromosome Location
Chr. 9: 21062393 - 21067093 on Build GRCm38
UniGene ID
Mm.483362
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001103168.1 NP_001096638.1 4, 5 687
GenBank mRNA DQ286956.1 - 4, 5 687

View Full Product Details