Gene Transcripts

Gene Symbol
Gpr25
Entrez Gene ID
383563
Gene Name
G protein-coupled receptor 25
Gene Aliases
Gm1300
Chromosome Location
Chr. 1: 136258914 - 136260873 on Build GRCm38
UniGene ID
Mm.40367
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101516.1 NP_001094986.1 1 901

View Full Product Details