Gene Transcripts

Gene Symbol
Krtap17-1
Entrez Gene ID
77914
Gene Name
keratin associated protein 17-1
Gene Aliases
A030006P16Rik
Chromosome Location
Chr. 11: 99993232 - 99993994 on Build GRCm38
UniGene ID
Mm.293692
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099774.2 NP_001093244.1 1 623
GenBank mRNA AK020698.1 - 1 624

View Full Product Details