Gene Transcripts

Gene Symbol
Adgrl2
Entrez Gene ID
99633
Gene Name
adhesion G protein-coupled receptor L2
Gene Aliases
AI450192, Gm619, Lec1, Lphh1, Lphn2, mKIAA0786
Chromosome Location
Chr. 3: 148815586 - 149445423 on Build GRCm38
UniGene ID
Mm.331434
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081298.1 NP_001074767.1 13, 14 2521
GenBank mRNA AK129215.1 - 6, 7 936

View Full Product Details