Gene Transcripts

Gene Symbol
Vmn2r96
Entrez Gene ID
433070
Gene Name
vomeronasal 2, receptor 96
Gene Aliases
EG433070
Chromosome Location
Chr. 17: 18573255 - 18598157 on Build GRCm38
UniGene ID
Mm.215962
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104547.1 NP_001098017.1 3 1105

View Full Product Details