Gene Transcripts
- Gene Symbol
- Sult2a6
- Entrez Gene ID
- 24902
- Gene Name
- sulfotransferase family 2A, dehydroepiandrosterone (DHEA)-preferring, member 6
- Gene Aliases
- RATSMP2A, RATSMP2B, Smp2a, St2a2, Sult2a4l1, Sult2al1
- Chromosome Location
- Chr. 1: 76529121 - 76722965 on Build Rnor_6.0
- UniGene ID
- Rn.221887
- Species
- Rattus norvegicus
- Gene Symbol
- LOC100360574
- Entrez Gene ID
- 100360574
- Gene Name
- alcohol sulfotransferase-like
- Gene Aliases
- Chromosome Location
- Chr. 1: 76004950 - 76053083 on Build Rnor_6.0
- Species
- Rattus norvegicus
- Gene Symbol
- Sult2a1
- Entrez Gene ID
- 24912
- Gene Name
- sulfotransferase family 2A, dehydroepiandrosterone (DHEA)-preferring, member 1
- Gene Aliases
- STa, Smp-2, St2, St2a1, Sth2
- Chromosome Location
- Chr. 1: 76558721 - 76614309 on Build Rnor_6.0
- UniGene ID
- Rn.91378
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_131903.1 |
NP_571978.1
|
2, 3 |
387 |
|
GenBank mRNA |
D14987.1 |
- |
2, 3 |
387 |
|
D14988.1 |
- |
2, 3 |
387 |
|
FQ209430.1 |
- |
2, 3 |
262 |
|
FQ209460.1 |
- |
2, 3 |
262 |
|
FQ210003.1 |
- |
2, 3 |
262 |
|
FQ218232.1 |
- |
2, 3 |
262 |
|
FQ218942.1 |
- |
2, 3 |
262 |
|
FQ218962.1 |
- |
2, 3 |
262 |
|
FQ219256.1 |
- |
2, 3 |
262 |
|
FQ219272.1 |
- |
2, 3 |
283 |
|
View Full Product Details