Gene Transcripts

Gene Symbol
Defb25
Entrez Gene ID
641644
Gene Name
defensin beta 25
Gene Aliases
Chromosome Location
Chr. 3: 148103643 - 148104483 on Build Rnor_6.0
UniGene ID
Rn.156440
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037516.1 NP_001032605.1 2 75
GenBank mRNA AY621356.1 - 2 75

View Full Product Details