Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014571.3 | NP_055386.1 | 2, 3 | 199 | |
GenBank mRNA | AF311885.1 | - | 2, 3 | 148 | |
AJ272215.1 | - | 2, 3 | 158 | ||
AK300463.1 | - | 2, 3 | 158 | ||
AL541019.3 | - | 2, 3 | 199 | ||
BC006087.1 | - | 2, 3 | 169 | 2964729 |