Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001317990.1 | NP_001304919.1 | 3 | 2346 | |
NM_015898.3 | NP_056982.1 | 3 | 2510 | ||
XM_005259570.4 | XP_005259627.3 | 3 | 3432 | ||
XM_005259571.4 | XP_005259628.1 | 3 | 3280 | ||
GenBank mRNA | BM696365.1 | - | 1 | 28 | |
BQ711712.1 | - | 321 |