Gene Transcripts
- Gene Symbol
- EPN3
- Entrez Gene ID
- 55040
- Gene Name
- epsin 3
- Gene Aliases
- Chromosome Location
- Chr. 17: 50532687 - 50543750 on Build GRCh38
- UniGene ID
- Hs.670090
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017957.2 |
NP_060427.2
|
10 |
2887 |
|
GenBank mRNA |
AK000785.1 |
- |
10 |
2836 |
|
View Full Product Details