Gene Transcripts

Gene Symbol
AGGF1
Entrez Gene ID
55109
Gene Name
angiogenic factor with G-patch and FHA domains 1
Gene Aliases
GPATC7, GPATCH7, HSU84971, HUS84971, VG5Q
Chromosome Location
Chr. 5: 77030385 - 77065234 on Build GRCh38
UniGene ID
Hs.634849
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018046.4 NP_060516.2 3 840
GenBank mRNA AW958325.1 - 3 322
AY500994.1 - 3 759
BC032844.1 - 3 826 5266978
BC073864.1 - 3 367
BE077189.1 - 1 115
U84971.1 - 3 769

View Full Product Details