Gene Transcripts

Gene Symbol
SLC29A3
Entrez Gene ID
55315
Gene Name
solute carrier family 29 member 3
Gene Aliases
ENT3, HCLAP, HJCD, PHID
Chromosome Location
Chr. 10: 71319253 - 71363390 on Build GRCh38
UniGene ID
Hs.438419
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001174098.1 NP_001167569.1 2 290
NM_018344.5 NP_060814.4 2 290
NR_033413.1 2 290
NR_033414.1 2 290
XM_006717910.3 XP_006717973.1 2 682
XM_017016377.1 XP_016871866.1 2 587
XM_017016378.1 XP_016871867.1 2 590
GenBank mRNA AF326987.1 - 1 238
AK002022.1 - 1 41
AK304503.1 - 2 260
AK314497.1 - 2 258
AK316152.1 - 2 275
AX775915.1 - 2 290
AX775917.1 - 2 290
AX775919.1 - 2 290
AX775921.1 - 2 290
AY288928.1 - 2 250
AY358686.1 - 2 268
BC041575.1 - 1 178
BC120996.1 - 2 268 40116513
BC120997.1 - 2 268 40116514
DC319920.1 - 2 290

View Full Product Details