Gene Transcripts

Gene Symbol
PLA2G2F
Entrez Gene ID
64600
Gene Name
phospholipase A2 group IIF
Gene Aliases
Chromosome Location
Chr. 1: 20139326 - 20150386 on Build GRCh38
UniGene ID
Hs.302034
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022819.3 NP_073730.3 5 606
XM_011541956.1 XP_011540258.1 5 610
GenBank mRNA AF306566.1 - 4 379
AK093645.1 - 5 610

View Full Product Details