Gene Transcripts
- Gene Symbol
- C12orf43
- Entrez Gene ID
- 64897
- Gene Name
- chromosome 12 open reading frame 43
- Gene Aliases
- Chromosome Location
- Chr. 12: 121002422 - 121016502 on Build GRCh38
- UniGene ID
- Hs.528671
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001286191.1 |
NP_001273120.1
|
2, 3 |
317 |
|
NM_001286192.1 |
NP_001273121.1
|
2, 3 |
224 |
|
NM_001286195.1 |
NP_001273124.1
|
2, 3 |
224 |
|
NM_001286196.1 |
NP_001273125.1
|
2, 3 |
224 |
|
NM_001286198.1 |
NP_001273127.1
|
3, 4 |
481 |
|
NM_022895.2 |
NP_075046.1
|
2, 3 |
224 |
|
NR_104409.1 |
|
2, 3 |
224 |
|
XM_017019829.1 |
XP_016875318.1
|
2, 3 |
308 |
|
GenBank mRNA |
AK022510.1 |
- |
2, 3 |
219 |
|
AK222630.1 |
- |
2, 3 |
209 |
|
AK225162.1 |
- |
2, 3 |
219 |
|
AK301566.1 |
- |
2, 3 |
209 |
|
AK307971.1 |
- |
2, 3 |
319 |
|
BC014661.1 |
- |
2, 3 |
198 |
4860511 |
BP201132.1 |
- |
2, 3 |
204 |
|
DA014862.1 |
- |
2, 3 |
213 |
|
DC297725.1 |
- |
2, 3 |
211 |
|
View Full Product Details