Gene Transcripts

Gene Symbol
COQ5
Entrez Gene ID
84274
Gene Name
coenzyme Q5, methyltransferase
Gene Aliases
Chromosome Location
Chr. 12: 120503279 - 120529171 on Build GRCh38
UniGene ID
Hs.17250
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032314.3 NP_115690.3 5, 6 796
XM_006719639.2 XP_006719702.1 6, 7 896
GenBank mRNA AK222610.1 - 5, 6 786
AK225242.1 - 5, 6 786
AK293656.1 - 5, 6 817
AK298214.1 - 4, 5 574
AK315681.1 - 5, 6 787
BC004916.2 - 5, 6 772
BC107874.1 - 5, 6 790 6452052
BX647562.1 - 5, 6 785
EU700459.1 - 5, 6 790

View Full Product Details