Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001014449.2 | NP_001014449.1 | 4 | 509 | |
NM_001014451.2 | NP_001014451.1 | 5 | 519 | ||
NM_001257172.1 | NP_001244101.1 | 5 | 444 | ||
NM_001257173.1 | NP_001244102.1 | 5 | 601 | ||
NM_001257174.1 | NP_001244103.1 | 4 | 555 | ||
NM_001257175.1 | NP_001244104.1 | 3 | 416 | ||
NM_007242.5 | NP_009173.1 | 6 | 612 | ||
XM_006721127.2 | XP_006721190.1 | 5 | 534 | ||
XM_011522832.2 | XP_011521134.1 | 7 | 668 | ||
XM_011522833.2 | XP_011521135.1 | 6 | 575 | ||
XM_011522834.2 | XP_011521136.1 | 6 | 557 | ||
XM_017022890.1 | XP_016878379.1 | 6 | 650 | ||
GenBank mRNA | AF353720.1 | - | 4 | 555 | |
AJ237946.1 | - | 6 | 473 | ||
AK027378.1 | - | 6 | 577 | ||
AK223115.1 | - | 6 | 583 | ||
AK296868.1 | - | 3 | 413 | ||
AK301938.1 | - | 5 | 601 | ||
AK302107.1 | - | 5 | 444 | ||
AK316346.1 | - | 4 | 490 | ||
AL136639.1 | - | 5 | 488 | ||
BC003626.2 | - | 6 | 576 | 3030038 | |
BC010008.2 | - | 4 | 461 | 4298615 | |
BC110805.1 | - | 5 | 383 | ||
BI756088.1 | - | 2 | 270 | ||
CR457215.1 | - | 6 | 473 | ||
CR533509.1 | - | 5 | 380 |