Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001293083.1 | NP_001280012.1 | 47 | 5316 | |
XM_011512110.2 | XP_011510412.1 | 47 | 5360 | ||
XM_011512111.2 | XP_011510413.1 | 47 | 5357 | ||
XM_011512113.2 | XP_011510415.1 | 47 | 5354 | ||
XM_011512114.2 | XP_011510416.1 | 47 | 5330 | ||
XM_011512115.2 | XP_011510417.1 | 46 | 5291 | ||
XM_011512116.2 | XP_011510418.1 | 47 | 5273 | ||
XM_011512117.2 | XP_011510419.1 | 46 | 5255 | ||
XM_011512118.2 | XP_011510420.1 | 46 | 5240 | ||
XM_011512119.2 | XP_011510421.1 | 46 | 5198 | ||
XM_011512120.2 | XP_011510422.1 | 46 | 5251 | ||
XM_011512122.2 | XP_011510424.1 | 47 | 5361 | ||
XM_011512124.2 | XP_011510426.1 | 31 | 4944 | ||
XM_017005222.1 | XP_016860711.1 | 47 | 5357 | ||
XM_017005223.1 | XP_016860712.1 | 30 | 4008 | ||
XM_017005224.1 | XP_016860713.1 | 21 | 2800 | ||
GenBank mRNA | AL133022.1 | - | 29 | 4719 | |
AY461813.1 | - | 46 | 5342 | ||
BC117324.1 | - | 13 | 1524 | ||
BC126368.1 | - | 13 | 1527 | ||
BC143825.1 | - | 13 | 1524 |