Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001004051.3 | NP_001004051.1 | 5 | 2362 | |
NM_001184874.2 | NP_001171803.1 | 5 | 2490 | ||
NM_001184875.2 | NP_001171804.1 | 4 | 2402 | ||
NM_001184876.2 | NP_001171805.1 | 5 | 2401 | ||
NM_138437.5 | NP_612446.1 | 4 | 2274 | ||
GenBank mRNA | AK094646.1 | - | 1 | 722 | |
AK096067.1 | - | 1 | 1610 | ||
AK123832.1 | - | 4 | 2274 | ||
AL832187.1 | - | 1 | 1002 | ||
BC013576.1 | - | 1 | 1606 | 3893954 | |
BC036772.1 | - | 4 | 2038 | 5723418 | |
BC051707.1 | - | 4 | 2752 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001199818.1 | NP_001186747.1 | 7 | 2494 |