Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024514.4 | NP_078790.2 | 5 | 1459 | |
XM_005252788.1 | XP_005252845.1 | 6 | 1749 | ||
XM_005252789.2 | XP_005252846.1 | 6 | 2002 | ||
XM_011519895.1 | XP_011518197.1 | 6 | 2153 | ||
XM_011519898.2 | XP_011518200.1 | 5 | 7783 | ||
XM_017017190.1 | XP_016872679.1 | 5 | 4344 | ||
XM_017017191.1 | XP_016872680.1 | 5 | 1964 | ||
XM_017017192.1 | XP_016872681.1 | 6 | 1949 | ||
XM_017017193.1 | XP_016872682.1 | 5 | 1911 | ||
XM_017017194.1 | XP_016872683.1 | 5 | 1764 | ||
XM_017017195.1 | XP_016872684.1 | 5 | 3507 | ||
GenBank mRNA | AY323817.1 | - | 5 | 1459 | |
BC104907.1 | - | 5 | 1504 | 8143910 | |
BC104909.1 | - | 5 | 1506 | 8143912 | |
BC111070.1 | - | 5 | 1508 |