Gene Transcripts

Gene Symbol
OR11H6
Entrez Gene ID
122748
Gene Name
olfactory receptor family 11 subfamily H member 6
Gene Aliases
Chromosome Location
Chr. 14: 20223710 - 20224702 on Build GRCh38
UniGene ID
Hs.553575
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004480.1 NP_001004480.1 1 532

View Full Product Details