Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001270396.1 | NP_001257325.1 | 2 | 428 | |
NM_152436.2 | NP_689649.1 | 2 | 428 | ||
NR_072995.1 | 3 | 558 | |||
XM_011537949.2 | XP_011536251.1 | 2 | 342 | ||
XM_011537950.2 | XP_011536252.1 | 2 | 314 | ||
XM_017018848.1 | XP_016874337.1 | 3 | 418 | ||
XM_017018849.1 | XP_016874338.1 | 3 | 444 | ||
GenBank mRNA | AK093175.1 | - | 2 | 418 | |
BC016749.1 | - | 2 | 435 | ||
BC029557.1 | - | 2 | 389 | 5262538 | |
BX640916.1 | - | 3 | 527 |