Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001409.3 | NP_001400.3 | 15 | 2047 | |
XM_006710406.3 | XP_006710469.1 | 12 | 1682 | ||
XM_011540885.1 | XP_011539187.1 | 16 | 2352 | ||
XM_011540886.1 | XP_011539188.1 | 16 | 2352 | ||
XM_011540887.2 | XP_011539189.1 | 11 | 1560 | ||
XM_011540888.2 | XP_011539190.1 | 16 | 2352 | ||
XM_017000533.1 | XP_016856022.1 | 15 | 2220 | ||
GenBank mRNA | AB011539.2 | - | 12 | 1708 | |
AB231860.1 | - | 15 | 2047 |