Gene Transcripts

Gene Symbol
KHDC1L
Entrez Gene ID
100129128
Gene Name
KH domain containing 1 like
Gene Aliases
Chromosome Location
Chr. 6: 73223544 - 73225452 on Build GRCh38
UniGene ID
Hs.663639
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001126063.2 NP_001119535.1 3 574

View Full Product Details