Gene Transcripts

Gene Symbol
ZSCAN23
Entrez Gene ID
222696
Gene Name
zinc finger and SCAN domain containing 23
Gene Aliases
ZNF390, ZNF453, dJ29K1.3, dJ29K1.3.1
Chromosome Location
Chr. 6: 28425516 - 28443554 on Build GRCh38
UniGene ID
Hs.721841
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012455.1 NP_001012458.1 4 2579
GenBank mRNA AL833347.1 - 4 2544

View Full Product Details