Gene Transcripts

Gene Symbol
GOLGA6A
Entrez Gene ID
342096
Gene Name
golgin A6 family member A
Gene Aliases
GLP, GOLGA6
Chromosome Location
Chr. 15: 74069857 - 74082550 on Build GRCh38
UniGene ID
Hs.546408
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001038640.2 NP_001033729.2 8 641
GenBank mRNA AK292574.1 - 8 622
BC131515.1 - 8 621

View Full Product Details