Gene Transcripts
- Gene Symbol
- USP17L26
- Entrez Gene ID
- 728379
- Gene Name
- ubiquitin specific peptidase 17-like family member 26
- Gene Aliases
- Chromosome Location
- Chr. 4: 9334658 - 9336250 on Build GRCh38
- UniGene ID
- Hs.553810
- Species
- Homo sapiens
- Gene Symbol
- USP17L1
- Entrez Gene ID
- 401447
- Gene Name
- ubiquitin specific peptidase 17-like family member 1
- Gene Aliases
- USP17L1P
- Chromosome Location
- Chr. 8: 7332387 - 7333979 on Build GRCh38
- UniGene ID
- Hs.653091
- Species
- Homo sapiens
- Gene Symbol
- USP17L10
- Entrez Gene ID
- 100287144
- Gene Name
- ubiquitin specific peptidase 17-like family member 10
- Gene Aliases
- Chromosome Location
- Chr. 4: 9210657 - 9212249 on Build GRCh38
- UniGene ID
- Hs.741130
- Species
- Homo sapiens
- Gene Symbol
- USP17L11
- Entrez Gene ID
- 100287178
- Gene Name
- ubiquitin specific peptidase 17-like family member 11
- Gene Aliases
- Chromosome Location
- Chr. 4: 9215405 - 9216997 on Build GRCh38
- UniGene ID
- Hs.722783
- Species
- Homo sapiens
- Gene Symbol
- USP17L12
- Entrez Gene ID
- 100287205
- Gene Name
- ubiquitin specific peptidase 17-like family member 12
- Gene Aliases
- Chromosome Location
- Chr. 4: 9220152 - 9221744 on Build GRCh38
- UniGene ID
- Hs.741128
- Species
- Homo sapiens
- Gene Symbol
- USP17L13
- Entrez Gene ID
- 100287238
- Gene Name
- ubiquitin specific peptidase 17-like family member 13
- Gene Aliases
- Chromosome Location
- Chr. 4: 9224896 - 9226488 on Build GRCh38
- UniGene ID
- Hs.741127
- Species
- Homo sapiens
- Gene Symbol
- USP17L15
- Entrez Gene ID
- 100288520
- Gene Name
- ubiquitin specific peptidase 17-like family member 15
- Gene Aliases
- Chromosome Location
- Chr. 4: 9234385 - 9236334 on Build GRCh38
- UniGene ID
- Hs.528621
- Species
- Homo sapiens
- Gene Symbol
- USP17L17
- Entrez Gene ID
- 100287327
- Gene Name
- ubiquitin specific peptidase 17-like family member 17
- Gene Aliases
- Chromosome Location
- Chr. 4: 9243879 - 9245471 on Build GRCh38
- UniGene ID
- Hs.741129
- Species
- Homo sapiens
- Gene Symbol
- USP17L18
- Entrez Gene ID
- 100287364
- Gene Name
- ubiquitin specific peptidase 17-like family member 18
- Gene Aliases
- Chromosome Location
- Chr. 4: 9248630 - 9250222 on Build GRCh38
- UniGene ID
- Hs.741138
- Species
- Homo sapiens
- Gene Symbol
- USP17L19
- Entrez Gene ID
- 100287404
- Gene Name
- ubiquitin specific peptidase 17-like family member 19
- Gene Aliases
- Chromosome Location
- Chr. 4: 9253378 - 9254970 on Build GRCh38
- UniGene ID
- Hs.741137
- Species
- Homo sapiens
- Gene Symbol
- USP17L2
- Entrez Gene ID
- 377630
- Gene Name
- ubiquitin specific peptidase 17-like family member 2
- Gene Aliases
- DUB-3, DUB3, USP17
- Chromosome Location
- Chr. 8: 12137168 - 12138760 on Build GRCh38
- UniGene ID
- Hs.531448
- Species
- Homo sapiens
- Gene Symbol
- USP17L20
- Entrez Gene ID
- 100287441
- Gene Name
- ubiquitin specific peptidase 17-like family member 20
- Gene Aliases
- Chromosome Location
- Chr. 4: 9258124 - 9259716 on Build GRCh38
- UniGene ID
- Hs.722783
- Species
- Homo sapiens
- Gene Symbol
- USP17L21
- Entrez Gene ID
- 100287478
- Gene Name
- ubiquitin specific peptidase 17-like family member 21
- Gene Aliases
- Chromosome Location
- Chr. 4: 9262872 - 9264464 on Build GRCh38
- UniGene ID
- Hs.741136
- Species
- Homo sapiens
- Gene Symbol
- USP17L22
- Entrez Gene ID
- 100287513
- Gene Name
- ubiquitin specific peptidase 17-like family member 22
- Gene Aliases
- Chromosome Location
- Chr. 4: 9267619 - 9269211 on Build GRCh38
- UniGene ID
- Hs.723053
- Species
- Homo sapiens
- Gene Symbol
- USP17L24
- Entrez Gene ID
- 728369
- Gene Name
- ubiquitin specific peptidase 17-like family member 24
- Gene Aliases
- Chromosome Location
- Chr. 4: 9325165 - 9326757 on Build GRCh38
- UniGene ID
- Hs.553810
- Species
- Homo sapiens
- Gene Symbol
- USP17L25
- Entrez Gene ID
- 728373
- Gene Name
- ubiquitin specific peptidase 17-like family member 25
- Gene Aliases
- Chromosome Location
- Chr. 4: 9329911 - 9331503 on Build GRCh38
- UniGene ID
- Hs.553810
- Species
- Homo sapiens
- Gene Symbol
- USP17L27
- Entrez Gene ID
- 728393
- Gene Name
- ubiquitin specific peptidase 17-like family member 27
- Gene Aliases
- Chromosome Location
- Chr. 4: 9344148 - 9345740 on Build GRCh38
- UniGene ID
- Hs.553810
- Species
- Homo sapiens
- Gene Symbol
- USP17L28
- Entrez Gene ID
- 728400
- Gene Name
- ubiquitin specific peptidase 17-like family member 28
- Gene Aliases
- Chromosome Location
- Chr. 4: 9348893 - 9350485 on Build GRCh38
- UniGene ID
- Hs.553810
- Species
- Homo sapiens
- Gene Symbol
- USP17L29
- Entrez Gene ID
- 728405
- Gene Name
- ubiquitin specific peptidase 17-like family member 29
- Gene Aliases
- Chromosome Location
- Chr. 4: 9353638 - 9355230 on Build GRCh38
- UniGene ID
- Hs.553810
- Species
- Homo sapiens
- Gene Symbol
- USP17L3
- Entrez Gene ID
- 645836
- Gene Name
- ubiquitin specific peptidase 17-like family member 3
- Gene Aliases
- USP17B, USP17F
- Chromosome Location
- Chr. 8: 7976393 - 7977985 on Build GRCh38
- UniGene ID
- Hs.741131
- Species
- Homo sapiens
- Gene Symbol
- USP17L30
- Entrez Gene ID
- 728419
- Gene Name
- ubiquitin specific peptidase 17-like family member 30
- Gene Aliases
- Chromosome Location
- Chr. 4: 9363129 - 9364721 on Build GRCh38
- UniGene ID
- Hs.553810
- Species
- Homo sapiens
- Gene Symbol
- USP17L4
- Entrez Gene ID
- 645402
- Gene Name
- ubiquitin specific peptidase 17-like family member 4
- Gene Aliases
- Chromosome Location
- Chr. 8: 7337115 - 7338707 on Build GRCh38
- UniGene ID
- Hs.741134
- Species
- Homo sapiens
- Gene Symbol
- USP17L5
- Entrez Gene ID
- 728386
- Gene Name
- ubiquitin specific peptidase 17-like family member 5
- Gene Aliases
- Chromosome Location
- Chr. 4: 9339403 - 9340995 on Build GRCh38
- UniGene ID
- Hs.740243
- Species
- Homo sapiens
- Gene Symbol
- USP17L6P
- Entrez Gene ID
- 391622
- Gene Name
- ubiquitin specific peptidase 17-like family member 6, pseudogene
- Gene Aliases
- DUB4, USP17L6, vDUB4
- Chromosome Location
- Chr. 4: 9367874 - 9369070 on Build GRCh38
- UniGene ID
- Hs.722658
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027279.1 |
|
1 |
250 |
|
GenBank mRNA |
AY533200.1 |
- |
1 |
250 |
|
- Gene Symbol
- USP17L9P
- Entrez Gene ID
- 391627
- Gene Name
- ubiquitin specific peptidase 17-like family member 9, pseudogene
- Gene Aliases
- RS447, USP17, USP17A, USP17H, USP17I, USP17J, USP17K, USP17L, USP17M
- Chromosome Location
- Chr. 4: 9358382 - 9359974 on Build GRCh38
- UniGene ID
- Hs.553810
- Species
- Homo sapiens
View Full Product Details