Gene Transcripts

Gene Symbol
ARGFX
Entrez Gene ID
503582
Gene Name
arginine-fifty homeobox
Gene Aliases
Chromosome Location
Chr. 3: 121567931 - 121590622 on Build GRCh38
UniGene ID
Hs.224976
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012659.1 NP_001012677.1 4 452

View Full Product Details