Gene Transcripts

Gene Symbol
ANAPC1
Entrez Gene ID
64682
Gene Name
anaphase promoting complex subunit 1
Gene Aliases
APC1, MCPR, TSG24
Chromosome Location
Chr. 2: 111766147 - 111884196 on Build GRCh38
UniGene ID
Hs.436527
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022662.3 NP_073153.1 38 4863
XM_011511633.2 XP_011509935.1 38 4907
XM_011511634.2 XP_011509936.1 38 5022
XM_017004710.1 XP_016860199.1 38 4907
XM_017004711.1 XP_016860200.1 38 4895
XM_017004712.1 XP_016860201.1 38 5010
XM_017004713.1 XP_016860202.1 37 4970
XM_017004714.1 XP_016860203.1 37 4906
GenBank mRNA AJ278357.1 - 38 4878
AK023807.1 - 25 3089
AL832013.1 - 27 3219
BC104902.1 - 38 4665 8143905
BC104904.1 - 38 4665 8143907

View Full Product Details