Gene Transcripts
- Gene Symbol
- ANKRD11
- Entrez Gene ID
- 29123
- Gene Name
- ankyrin repeat domain 11
- Gene Aliases
- ANCO-1, ANCO1, LZ16, T13
- Chromosome Location
- Chr. 16: 89267619 - 89490561 on Build GRCh38
- UniGene ID
- Hs.335003
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001256182.1 |
NP_001243111.1
|
12, 13 |
8253 |
|
NM_001256183.1 |
NP_001243112.1
|
11, 12 |
8179 |
|
NM_013275.5 |
NP_037407.4
|
11, 12 |
8182 |
|
XM_006721181.1 |
XP_006721244.1
|
11, 12 |
7959 |
|
XM_006721184.2 |
XP_006721247.1
|
10, 11 |
7997 |
|
XM_011523051.2 |
XP_011521353.1
|
12, 13 |
8242 |
|
XM_011523053.2 |
XP_011521355.1
|
12, 13 |
8348 |
|
XM_011523054.2 |
XP_011521356.1
|
12, 13 |
8262 |
|
XM_011523055.2 |
XP_011521357.1
|
13, 14 |
8547 |
|
XM_011523056.2 |
XP_011521358.1
|
12, 13 |
8284 |
|
XM_011523057.2 |
XP_011521359.1
|
11, 12 |
8165 |
|
XM_017023182.1 |
XP_016878671.1
|
11, 12 |
8615 |
|
XM_017023183.1 |
XP_016878672.1
|
12, 13 |
8351 |
|
XM_017023184.1 |
XP_016878673.1
|
12, 13 |
8348 |
|
XM_017023185.1 |
XP_016878674.1
|
10, 11 |
7989 |
|
XM_017023186.1 |
XP_016878675.1
|
14, 15 |
8725 |
|
XM_017023187.1 |
XP_016878676.1
|
10, 11 |
8077 |
|
GenBank mRNA |
AY373756.1 |
- |
11, 12 |
8185 |
|
AY533563.1 |
- |
12, 13 |
8010 |
|
KJ534770.1 |
- |
7, 8 |
995 |
|
View Full Product Details