Gene Transcripts

Gene Symbol
ANKRD11
Entrez Gene ID
29123
Gene Name
ankyrin repeat domain 11
Gene Aliases
ANCO-1, ANCO1, LZ16, T13
Chromosome Location
Chr. 16: 89267619 - 89490561 on Build GRCh38
UniGene ID
Hs.335003
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256182.1 NP_001243111.1 13 8316
NM_001256183.1 NP_001243112.1 12 8242
NM_013275.5 NP_037407.4 12 8245
XM_006721181.1 XP_006721244.1 12 8022
XM_006721184.2 XP_006721247.1 11 8060
XM_011523051.2 XP_011521353.1 13 8305
XM_011523053.2 XP_011521355.1 13 8411
XM_011523054.2 XP_011521356.1 13 8325
XM_011523055.2 XP_011521357.1 14 8610
XM_011523056.2 XP_011521358.1 13 8347
XM_011523057.2 XP_011521359.1 12 8228
XM_017023182.1 XP_016878671.1 12 8678
XM_017023183.1 XP_016878672.1 13 8414
XM_017023184.1 XP_016878673.1 13 8411
XM_017023185.1 XP_016878674.1 11 8052
XM_017023186.1 XP_016878675.1 15 8788
XM_017023187.1 XP_016878676.1 11 8140
GenBank mRNA AY373756.1 - 12 8248
AY533563.1 - 13 8073
KJ534770.1 - 8 1058

View Full Product Details