Gene Transcripts

Gene Symbol
CUEDC1
Entrez Gene ID
404093
Gene Name
CUE domain containing 1
Gene Aliases
Chromosome Location
Chr. 17: 57861243 - 57955323 on Build GRCh38
UniGene ID
Hs.46679
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001292025.1 NP_001278954.1 1 392
GenBank mRNA AK000746.1 - 1 392

View Full Product Details