Gene Transcripts
- Gene Symbol
- UBA1
- Entrez Gene ID
- 7317
- Gene Name
- ubiquitin like modifier activating enzyme 1
- Gene Aliases
- A1S9, A1S9T, A1ST, AMCX1, CFAP124, GXP1, POC20, SMAX2, UBA1A, UBE1, UBE1X
- Chromosome Location
- Chr. X: 47190800 - 47215128 on Build GRCh38
- UniGene ID
- Hs.533273
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003334.3 |
NP_003325.2
|
23, 24 |
3058 |
|
NM_153280.2 |
NP_695012.1
|
23, 24 |
2986 |
|
XM_005272649.1 |
XP_005272706.1
|
24, 25 |
3436 |
|
XM_011543954.2 |
XP_011542256.1
|
24, 25 |
3078 |
|
XM_017029777.1 |
XP_016885266.1
|
23, 24 |
4153 |
|
XM_017029778.1 |
XP_016885267.1
|
24, 25 |
5327 |
|
XM_017029779.1 |
XP_016885268.1
|
23, 24 |
5835 |
|
XM_017029780.1 |
XP_016885269.1
|
23, 24 |
2943 |
|
XM_017029781.1 |
XP_016885270.1
|
23, 24 |
2991 |
|
GenBank mRNA |
AK097343.1 |
- |
7, 8 |
1958 |
|
AK131341.1 |
- |
2, 3 |
1182 |
|
AK293161.1 |
- |
12, 13 |
1650 |
|
AK296866.1 |
- |
13, 14 |
1676 |
|
BC013041.2 |
- |
23, 24 |
2925 |
3543021 |
BC020261.1 |
- |
20, 21 |
2620 |
|
M58028.1 |
- |
23, 24 |
2964 |
|
X52897.1 |
- |
16, 17 |
2130 |
|
X56976.1 |
- |
23, 24 |
2867 |
|
View Full Product Details